5-Bromo-1-methylpyrimidin-2(1H)-one 95%
Lieferant: Apollo Scientific
OR928790-1G
APOSOR928790-1GEA
74.1
EUR
APOSOR928790-1G
5-Bromo-1-methylpyrimidin-2(1H)-one 95%
5-Bromo-1-methylpyrimidin-2(1H)-one
Formel:
C₅H₅BrN₂O Molecular Weight: 189.01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044136 CAS-Nummer: 14248-01-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...