Butyl methanesulphonate 95%

Lieferant: Apollo Scientific

OR928832-10G OR928832-5G OR928832-25G
APOSOR928832-10GEA 82.8 EUR
APOSOR928832-10G APOSOR928832-5G APOSOR928832-25G
Butyl methanesulphonate 95%
Butyl methanesulphonate
Formel: C₅H₁₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00041257
CAS-Nummer: 1912-32-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR