3-(2-Nitrophenyl)-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR9289-1G OR9289-250MG
APOSOR9289-1GEA 163 EUR
APOSOR9289-1G APOSOR9289-250MG
3-(2-Nitrophenyl)-1H-pyrazole
3-(2-Nitrophenyl)-1H-pyrazole
Formel: C₉H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00665858
CAS-Nummer: 59844-05-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR