2-(2-Chlorethoxy)ethanol 97%

Lieferant: Apollo Scientific

OR928909-500G
APOSOR928909-500GEA 68.3 EUR
APOSOR928909-500G
2-(2-Chlorethoxy)ethanol 97%
2-(2-Chloroethoxy)ethanol
Formel: C₄H₉ClO₂
Molecular Weight: 124.57 g/mol
Siedepunkt: 79
Lagertemperatur: Raumtemperatur
MDL: MFCD00002870
CAS-Nummer: 628-89-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR