Methyl-2-bromo-5-(bromomethyl)benzoate 95%

Lieferant: Apollo Scientific
Achtung

OR928929-1G OR928929-250MG OR928929-5G
APOSOR928929-1GEA 313 EUR
APOSOR928929-1G APOSOR928929-250MG APOSOR928929-5G
Methyl-2-bromo-5-(bromomethyl)benzoate 95%
Methyl-2-bromo-5-(bromomethyl)benzoate
Formel: C₉H₈Br₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD18398708
CAS-Nummer: 90721-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR