1-(4-Isocyanatobenzyl)piperidine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-Isocyanato-4-[(piperidin-1-yl)methyl]benzene, 4-[(Piperidin-1-yl)methyl]phenyl isocyanate

OR9290-50MG
APOSOR9290-50MGEA 36.4 EUR
APOSOR9290-50MG
1-(4-Isocyanatobenzyl)piperidine
1-(4-Isocyanatobenzyl)piperidine

May form small amount of sediment

Formel: C₁₃H₁₆N₂O
Molecular Weight: 216,279 g/mol
Siedepunkt: 85…90 °C (0,1 mm Hg)
Lagertemperatur: Kühlschrank
MDL: MFCD08435873
CAS-Nummer: 879896-46-5
UN: 2206
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR