1-Furan-3-yl-ethylamine 96%

Lieferant: Apollo Scientific

OR929031-1G OR929031-250MG
APOSOR929031-1GEA 476 EUR
APOSOR929031-1G APOSOR929031-250MG
1-Furan-3-yl-ethylamine 96%
1-Furan-3-yl-ethylamine
Formel: C₆H₉NO
Molecular Weight: 111.142 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06738794
CAS-Nummer: 252372-09-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR