(2-Ethoxyphenyl)essigsäure 95%

Lieferant: Apollo Scientific

OR929038-5G
APOSOR929038-5GEA 112 EUR
APOSOR929038-5G
(2-Ethoxyphenyl)essigsäure 95%
(2-Ethoxyphenyl)essigsäure
Formel: H₃CCH₂O(C₆H₄)CH₂COOH
Lagertemperatur: Raumtemperatur
MDL: MFCD00016824
CAS-Nummer: 70289-12-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR