Cyclopropyl(3-pyridinyl)methanone 95%

Lieferant: Apollo Scientific

OR929114-1G
APOSOR929114-1GEA 82.8 EUR
APOSOR929114-1G
Cyclopropyl(3-pyridinyl)methanone 95%
Cyclopropyl(3-pyridinyl)methanone
Formel: C₉H₉NO
Molecular Weight: 147,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864467
CAS-Nummer: 24966-13-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR