5-Bromo-L-tryptophan 97%

Lieferant: Apollo Scientific

OR929180-1G OR929180-250MG OR929180-500MG OR929180-100MG
APOSOR929180-1GEA 0 EUR
APOSOR929180-1G APOSOR929180-250MG APOSOR929180-500MG APOSOR929180-100MG
5-Bromo-L-tryptophan 97%
5-Bromo-L-tryptophan
Formel: C₁₁H₁₁BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06796707
CAS-Nummer: 25197-99-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR