Ac-Ile-OMe 95%

Lieferant: Apollo Scientific

OR929225-1G OR929225-5G
APOSOR929225-1GEA 0 EUR
APOSOR929225-1G APOSOR929225-5G
Ac-Ile-OMe 95%
Ac-Ile-OMe
Formel: C₉H₁₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00080833
CAS-Nummer: 2256-76-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR