3-Chlor-4-nitrophenol 98%

Lieferant: Apollo Scientific

OR929226-10G
APOSOR929226-10GEA 87 EUR
APOSOR929226-10G
3-Chlor-4-nitrophenol 98%
3-Chlor-4-nitrophenol
Formel: C₆H₄ClNO₃
Molecular Weight: 173.56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00466090
CAS-Nummer: 491-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR