2-(T-Butyloxy)-ethylamine hydrochloride 95%

Lieferant: Apollo Scientific

OR929232-1G
APOSOR929232-1GEA 270 EUR
APOSOR929232-1G
2-(T-Butyloxy)-ethylamine hydrochloride 95%
2-(T-Butyloxy)-ethylamine hydrochloride
Formel: C₆H₁₅NO·ClH
Molecular Weight: 153,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973191
CAS-Nummer: 335598-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR