Ethyl-3-oxocyclopentane-1-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-oxocyclopentanecarboxylate

OR929246-5G
APOSOR929246-5GEA 59.6 EUR
APOSOR929246-5G
Ethyl-3-oxocyclopentane-1-carboxylate 97%
Ethyl-3-oxocyclopentane-1-carboxylate
Formel: C₈H₁₂O₃
Molecular Weight: 156.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01320352
CAS-Nummer: 5400-79-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR