5,5-Dimethylpyrrolidin-2-one 95%

Lieferant: Apollo Scientific

OR929277-5G
APOSOR929277-5GEA 112 EUR
APOSOR929277-5G
5,5-Dimethylpyrrolidin-2-one 95%
5,5-Dimethylpyrrolidin-2-one
Formel: C₆H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00128876
CAS-Nummer: 5165-28-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR