Methyl 2-[(dimethylamino)methyl]benzoate 95%
Lieferant: Apollo Scientific
OR929284-1G
OR929284-250MG
APOSOR929284-1GEA
538
EUR
APOSOR929284-1G
APOSOR929284-250MG
Methyl 2-[(dimethylamino)methyl]benzoate 95%
Methyl 2-[(dimethylamino)methyl]benzoate
Formel:
C₁₁H₁₅NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13249350 CAS-Nummer: 27171-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...