2-(2,4-Dimethylphenoxy)acetohydrazide

Lieferant: Apollo Scientific
Achtung

OR9293-100G
APOSOR9293-100GEA 174 EUR
APOSOR9293-100G
2-(2,4-Dimethylphenoxy)acetohydrazide
2-(2,4-Dimethylphenoxy)acetohydrazide
Formel: C₁₀H₁₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00156144
CAS-Nummer: 72293-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR