3-Chloro-4-ethoxybenzaldehyde 95%

Lieferant: Apollo Scientific

OR929320-1G OR929320-5G
APOSOR929320-1GEA 88.6 EUR
APOSOR929320-1G APOSOR929320-5G
3-Chloro-4-ethoxybenzaldehyde 95%
3-Chloro-4-ethoxybenzaldehyde
Formel: C₉H₉ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06246171
CAS-Nummer: 99585-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR