trans-2-Octenoic acid 95%

Lieferant: Apollo Scientific

OR929368-5G
APOSOR929368-5GEA 0 EUR
APOSOR929368-5G
trans-2-Octenoic acid 95%
trans-2-Octenoic acid
Formel: C₈H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00002706
CAS-Nummer: 1871-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR