3,3'-Bipyridin 95%
Lieferant: Apollo Scientific
Synonyme:
3,3'-Dipyridyl, 3,3'-Bipyridine
OR929371-1G
APOSOR929371-1GEA
159
EUR
APOSOR929371-1G
3,3'-Bipyridin 95%
3,3'-Bipyridin
Formel:
C₁₀H₈N₂ Molecular Weight: 156,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006371 CAS-Nummer: 581-46-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...