2,6-Dimethylphenylisocyanat 95%
Lieferant: Apollo Scientific
Synonyme:
2,6-Xylylisocyanat
OR929424-25G
APOSOR929424-25GEA
149
EUR
APOSOR929424-25G
2,6-Dimethylphenylisocyanat 95%
2,6-Dimethylphenylisocyanat
- May form small amount of sediment.
Formel:
C₉H₉NO Molecular Weight: 147.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002012 CAS-Nummer: 28556-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...