Ethyl-N-methyl-β-alaninate 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-(methylamino)propanoate

OR929512-10G OR929512-1G OR929512-25G
APOSOR929512-10GEA 171 EUR
APOSOR929512-10G APOSOR929512-1G APOSOR929512-25G
Ethyl-N-methyl-β-alaninate 98%
Ethyl-N-methyl-beta-alaninate
Formel: C₆H₁₃NO₂
Molecular Weight: 131.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672194
CAS-Nummer: 2213-08-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR