2,4-Dimethylbenzoxazole 95%

Lieferant: Apollo Scientific

OR929525-1G
APOSOR929525-1GEA 364 EUR
APOSOR929525-1G
2,4-Dimethylbenzoxazole 95%
2,4-Dimethylbenzoxazole
Formel: C₉H₉NO
Molecular Weight: 147,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21641329
CAS-Nummer: 72692-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR