3,6,6-Trimethyl-1,5,6,7-tetrahydro-4h-indazol-4-one 95%

Lieferant: Apollo Scientific

OR929611-1G
APOSOR929611-1GEA 574 EUR
APOSOR929611-1G
3,6,6-Trimethyl-1,5,6,7-tetrahydro-4h-indazol-4-one 95%
3,6,6-Trimethyl-1,5,6,7-tetrahydro-4h-indazol-4-one
Formel: C₁₀H₁₄N₂O
Molecular Weight: 178,231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00474387
CAS-Nummer: 16315-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR