(2,4-Dimethylphenyl)essigsäure 98%

Lieferant: Apollo Scientific

Synonyme: 2,4-Xylylessigsäure

OR929615-100G OR929615-10G OR929615-25G
APOSOR929615-100GEA 601 EUR
APOSOR929615-100G APOSOR929615-10G APOSOR929615-25G
(2,4-Dimethylphenyl)essigsäure 98%
(2,4-Dimethylphenyl)essigsäure
Formel: (C₆H₃(CH₃)₂)CH₂COOH
Lagertemperatur: Raumtemperatur
MDL: MFCD02664685
CAS-Nummer: 6331-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR