7-Methoxy-3,4-dihydro-2H-isoquinolin-1-one 95%

Lieferant: Apollo Scientific

Synonyme: 7-Methoxy-3,4-dihydroisoquinolin-1(2H)-one

OR929617-1G
APOSOR929617-1GEA 116 EUR
APOSOR929617-1G
7-Methoxy-3,4-dihydro-2H-isoquinolin-1-one 95%
7-Methoxy-3,4-dihydro-2H-isoquinolin-1-one
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114863
CAS-Nummer: 22246-04-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR