5-(2-Chloroethyl)-1,3-benzodioxole 97%

Lieferant: Apollo Scientific

Synonyme: 4-(2-Chloroethyl)-1,2-methylenedioxybenzene

OR9297-1G
APOSOR9297-1GEA 183 EUR
APOSOR9297-1G
5-(2-Chloroethyl)-1,3-benzodioxole 97%
5-(2-Chloroethyl)-1,3-benzodioxole
Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
Siedepunkt: 112 °C (2 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD08435910
CAS-Nummer: 23808-46-0
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR