1-(2-(Methylamino)phenyl)ethanone 95%

Lieferant: Apollo Scientific

OR929725-1G
APOSOR929725-1GEA 177 EUR
APOSOR929725-1G
1-(2-(Methylamino)phenyl)ethanone 95%
1-(2-(Methylamino)phenyl)ethanone
Formel: C₉H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD19159692
CAS-Nummer: 1859-75-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR