4-Chloro-1-phenylpyrazole 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-1-phenyl-1H-pyrazole

OR929752-100MG
APOSOR929752-100MGEA 78.3 EUR
APOSOR929752-100MG
4-Chloro-1-phenylpyrazole 95%
4-Chloro-1-phenylpyrazole
Formel: C₉H₇ClN₂
Molecular Weight: 178,618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234512
CAS-Nummer: 6831-92-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR