N,N-Diethyl-1,4-cyclohexanediamine (cis and trans mixture) 95%

Lieferant: Apollo Scientific

OR929777-1G
APOSOR929777-1GEA 78.3 EUR
APOSOR929777-1G
N,N-Diethyl-1,4-cyclohexanediamine (cis and trans mixture) 95%
N,N-Diethyl-1,4-cyclohexanediamine (cis and trans mixture)
Formel: C₁₀H₂₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06658428
CAS-Nummer: 42389-54-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR