4-Chloro-2-methyl-6-(methylthio)pyrimidine 95%

Lieferant: Apollo Scientific

OR929794-1G
APOSOR929794-1GEA 163 EUR
APOSOR929794-1G
4-Chloro-2-methyl-6-(methylthio)pyrimidine 95%
4-Chloro-2-methyl-6-(methylthio)pyrimidine
Formel: C₆H₇ClN₂S
Molecular Weight: 174.65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16769133
CAS-Nummer: 867131-59-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR