1-Methyl-piperidine-4-carbonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 1-Methylpiperidine-4-carbonitrile

OR929855-1G
APOSOR929855-1GEA 104 EUR
APOSOR929855-1G
1-Methyl-piperidine-4-carbonitrile 95%
1-Methyl-piperidine-4-carbonitrile
Formel: C₇H₁₂N₂
Molecular Weight: 124.184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658487
CAS-Nummer: 20691-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR