Z-DL-ALA-GLY-OH

Lieferant: Apollo Scientific
Achtung

OR929864-1G
APOSOR929864-1GEA 161 EUR
APOSOR929864-1G
Z-DL-ALA-GLY-OH
Z-DL-ALA-GLY-OH
Formel: C₁₃H₁₆N₂O₅
Molecular Weight: 280,277 g/mol
Schmelzpunkt: 130…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00021731
CAS-Nummer: 2503-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR