2,4-Dinitro-1-naphthol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Acid Yellow 24, 1,3-Dinitro-4-hydroxynaphthalin, Martiumgelb, Naphtholgelb, Martinsgelb, 2,4-Dinitronaphthol

OR929998-25G
APOSOR929998-25GEA 183 EUR
APOSOR929998-25G
2,4-Dinitro-1-naphthol 98%
2,4-Dinitro-1-naphthol
Formel: C₁₀H₆N₂O₅
Molecular Weight: 234,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00065352
CAS-Nummer: 605-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR