Methyl-4-chlor-3-nitrobenzoat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chlor-3-nitrobenzoesäuremethylester
OR9300-5G
APOSOR9300-5GEA
21.9
EUR
APOSOR9300-5G
Methyl-4-chlor-3-nitrobenzoat 98%
Methyl-4-chlor-3-nitrobenzoat
Formel:
C₈H₆ClNO₄ Molecular Weight: 215,59 g/mol Schmelzpunkt: 80…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016989 CAS-Nummer: 14719-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...