7-Bromothieno[3,2-d]pyrimidine 98%

Lieferant: Apollo Scientific
Achtung

OR930033-250MG
APOSOR930033-250MGEA 187 EUR
APOSOR930033-250MG
7-Bromothieno[3,2-d]pyrimidine 98%
7-Bromothieno[3,2-d]pyrimidine
Formel: C₆H₃BrN₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD11520387
CAS-Nummer: 21586-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR