Methyl-2-hydroxy-3-phenylpropanoate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-hydroxy-3-phenylpropanoate

OR930185-10G OR930185-1G OR930185-5G
APOSOR930185-10GEA 116 EUR
APOSOR930185-10G APOSOR930185-1G APOSOR930185-5G
Methyl-2-hydroxy-3-phenylpropanoate 95%
Methyl-2-hydroxy-3-phenylpropanoate
Formel: C₁₀H₁₂O₃
Molecular Weight: 180.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06654233
CAS-Nummer: 13674-16-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR