M-Aminobenzyl cyanide 95%

Lieferant: Apollo Scientific

Synonyme: 2-(3-Aminophenyl)acetonitrile

OR930194-1G
APOSOR930194-1GEA 105 EUR
APOSOR930194-1G
M-Aminobenzyl cyanide 95%
M-Aminobenzyl cyanide
Formel: C₈H₈N₂
Molecular Weight: 132,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03265299
CAS-Nummer: 4623-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR