(1-Fmoc-aminocyclohexyl)acetic acid 97%

Lieferant: Apollo Scientific

OR930222-1G
APOSOR930222-1GEA 319 EUR
APOSOR930222-1G
(1-Fmoc-aminocyclohexyl)acetic acid 97%
(1-Fmoc-aminocyclohexyl)acetic acid
Formel: C₂₃H₂₅NO₄
Molecular Weight: 379.449 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01863220
CAS-Nummer: 282524-98-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR