(2,2,6,6-Tetramethyl-4-oxo-1-piperidinyl)oxidanyl 95%
Lieferant: Apollo Scientific
OR930328-25G
APOSOR930328-25GEA
78.3
EUR
APOSOR930328-25G
(2,2,6,6-Tetramethyl-4-oxo-1-piperidinyl)oxidanyl 95%
(2,2,6,6-Tetramethyl-4-oxo-1-piperidinyl)oxidanyl
Formel:
C₉H₁₆NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006187 CAS-Nummer: 2896-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...