2,3,5-Trichlorphenol 95%
Lieferant: Apollo Scientific
OR930342-5G
APOSOR930342-5GEA
183
EUR
APOSOR930342-5G
2,3,5-Trichlorphenol 95%
2,3,5-Trichlorphenol
Formel:
C₆H₃Cl₃O Molecular Weight: 197.45 g/mol Siedepunkt: 248 Schmelzpunkt: 62 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002164 CAS-Nummer: 933-78-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...