2-Chlor-4-phenylpyrimidin 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR9304-1G
APOSOR9304-1GEA
33.5
EUR
APOSOR9304-1G
2-Chlor-4-phenylpyrimidin 95%
2-Chloro-4-phenylpyrimidine
Formel:
C₁₀H₇ClN₂ Molecular Weight: 190.63 g/mol Schmelzpunkt: 84…88 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00234600 CAS-Nummer: 13036-50-5 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...