Methyl-p-benzochinon 98%

Lieferant: Apollo Scientific

Synonyme: Methyl-1,4-benzochinon, Methylchinon

OR930410-500G
APOSOR930410-500GEA 177 EUR
APOSOR930410-500G
Methyl-p-benzochinon 98%
Methyl-p-benzochinon
Formel: C₇H₆O₂
Molecular Weight: 122.12 g/mol
Siedepunkt: 187
Lagertemperatur: Raumtemperatur
MDL: MFCD00001603
CAS-Nummer: 553-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR