5,7-Dimethylpyrazolo[1,5-a]pyrimidine 95%

Lieferant: Apollo Scientific

OR930461-1G
APOSOR930461-1GEA 435 EUR
APOSOR930461-1G
5,7-Dimethylpyrazolo[1,5-a]pyrimidine 95%
5,7-Dimethylpyrazolo[1,5-a]pyrimidine
Formel: C₈H₉N₃
Molecular Weight: 147,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01692984
CAS-Nummer: 35149-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR