Methyl 2-(cyclopentylamino)acetate hydrochloride 95%

Lieferant: Apollo Scientific

OR930493-5G
APOSOR930493-5GEA 126 EUR
APOSOR930493-5G
Methyl 2-(cyclopentylamino)acetate hydrochloride 95%
Methyl 2-(cyclopentylamino)acetate hydrochloride
Formel: C₈H₁₅NO₂·ClH
Molecular Weight: 193,671 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15071418
CAS-Nummer: 195877-46-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR