1-Phenyl-2-pyrrolidinon 98%

Lieferant: Apollo Scientific

Synonyme: 1-Phenyl-2-pyrrolidinone

OR930513-100G
APOSOR930513-100GEA 85.7 EUR
APOSOR930513-100G
1-Phenyl-2-pyrrolidinon 98%
1-Phenyl-2-pyrrolidone
Formel: C₁₀H₁₁NO
Molecular Weight: 161.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003192
CAS-Nummer: 4641-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR