3-Cyclohexyl-3-oxopropanenitrile 95%

Lieferant: Apollo Scientific

OR930579-1G OR930579-250MG
APOSOR930579-1GEA 806 EUR
APOSOR930579-1G APOSOR930579-250MG
3-Cyclohexyl-3-oxopropanenitrile 95%
3-Cyclohexyl-3-oxopropanenitrile
Formel: C₉H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD07700128
CAS-Nummer: 62455-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR