4-Hydroxy-8-methyl-2(1H)-quinolinone 95%
Lieferant: Apollo Scientific
OR930675-1G
APOSOR930675-1GEA
487
EUR
APOSOR930675-1G
4-Hydroxy-8-methyl-2(1H)-quinolinone 95%
4-Hydroxy-8-methyl-2(1H)-quinolinone
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,184 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01108967 CAS-Nummer: 1677-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...