4-Methyl-2-nitrobenzaldehyde 95%

Lieferant: Apollo Scientific

OR930693-1G
APOSOR930693-1GEA 94.4 EUR
APOSOR930693-1G
4-Methyl-2-nitrobenzaldehyde 95%
4-Methyl-2-nitrobenzaldehyde
Formel: C₈H₇NO₃
Molecular Weight: 165.146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669895
CAS-Nummer: 20357-22-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR