4-(2-Methoxyphenyl)pyrrolidin-2-one 95%

Lieferant: Apollo Scientific

OR930701-1G
APOSOR930701-1GEA 609 EUR
APOSOR930701-1G
4-(2-Methoxyphenyl)pyrrolidin-2-one 95%
4-(2-Methoxyphenyl)pyrrolidin-2-one
Formel: C₁₁H₁₃NO₂
Molecular Weight: 191,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09955050
CAS-Nummer: 1020718-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR